Learn More
MSC Value Collection Plug Gage Set, 0.917-1 in, 84 pc
Catalog No. 5000529725
Encompass
This Class ZZ Plug Gage Set provides precise measurements and offers great value for various business needs. Designed for accuracy, it ensures reliable performance in demanding applications.
  • Provides a measuring range of 0.917-1 in.
  • Includes 84 individual pieces for comprehensive gaging.
  • Features a minimum diameter of 0.9170 decimal "
  • Features a maximum diameter of 1.0000 decimal "
  • Maintains a tight tolerance of 0.000200 in.
  • Designed with a plus tolerance type for specific applications.
  • Classified as Class ZZ for precision.
Catalog No. 50-005-29725 Supplier MSC Supplier No. 05046693
This Class ZZ Plug Gage Set provides precise measurements and offers great value for various business needs. Designed for accuracy, it ensures reliable performance in demanding applications.
  • Provides a measuring range of 0.917-1 in.
  • Includes 84 individual pieces for comprehensive gaging.
  • Features a minimum diameter of 0.9170 decimal "
  • Features a maximum diameter of 1.0000 decimal "
  • Maintains a tight tolerance of 0.000200 in.
  • Designed with a plus tolerance type for specific applications.
  • Classified as Class ZZ for precision.