Learn More
MSC Value Collection Plug Gage Set, Class ZZ, 0.251-0.5 in Range, 250 Pc
Catalog No. 5000514213
Encompass
This Class ZZ plug gage set is designed for precise measurement and inspection tasks. It covers a diameter range from 0.251 to 0.5 inches with a tolerance of 0.000200 inch, and includes 250 pieces made from durable steel.
  • Includes 250 steel pieces for comprehensive gaging needs.
  • Features a 0.000200 inch tolerance for high precision measurements.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Enables NO-GO gaging inspection.
  • Assists in gaging slot widths and locations.
  • Allows for accurate measurement of hole diameters.
  • Useful for calculating distances between holes.
Catalog No. 50-005-14213 Supplier MSC Supplier No. 86463023
This Class ZZ plug gage set is designed for precise measurement and inspection tasks. It covers a diameter range from 0.251 to 0.5 inches with a tolerance of 0.000200 inch, and includes 250 pieces made from durable steel.
  • Includes 250 steel pieces for comprehensive gaging needs.
  • Features a 0.000200 inch tolerance for high precision measurements.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Enables NO-GO gaging inspection.
  • Assists in gaging slot widths and locations.
  • Allows for accurate measurement of hole diameters.
  • Useful for calculating distances between holes.