Learn More
MSC Value Collection Plug Gage Set, Class ZZ, 0.917-1 in, 84 Pc
Catalog No. 5000534540
Encompass
This Class ZZ plug gage set provides a range of 0.917 to 1 inch with a tolerance of 0.000200 " It includes 84 pieces and offers a cost-effective solution for precision measurement needs.
  • Minimum diameter of 0.9170 inch
  • Maximum diameter of 1.0000 inch
  • Tolerance of 0.000200 inch
  • Class ZZ for precise measurements
  • Minus tolerance type
  • Includes 84 pieces
Catalog No. 50-005-34540 Supplier MSC Supplier No. 86463387
This Class ZZ plug gage set provides a range of 0.917 to 1 inch with a tolerance of 0.000200 " It includes 84 pieces and offers a cost-effective solution for precision measurement needs.
  • Minimum diameter of 0.9170 inch
  • Maximum diameter of 1.0000 inch
  • Tolerance of 0.000200 inch
  • Class ZZ for precise measurements
  • Minus tolerance type
  • Includes 84 pieces