Learn More
MSC Value Collection Polybag, 12 x 15 in, 4 mil, 500/Pk
Catalog No. 5000794479
Encompass
These reclosable polybags are manufactured from 100% virgin, high clarity polyethylene film. Designed for heavy-duty applications, they are ideal for storing industrial hardware and tools, keeping contents clean and protected from moisture.
  • Zipper closure ensures secure sealing and reusability
  • Manufactured from high clarity polyethylene film
  • Meets FDA and USDA specifications for safe contact
  • Thickness of 4 mil provides durability for demanding applications
  • Inside usable dimensions are 12 in W x 15 in H
Catalog No. 50-007-94479 Supplier MSC Supplier No. 46056271
These reclosable polybags are manufactured from 100% virgin, high clarity polyethylene film. Designed for heavy-duty applications, they are ideal for storing industrial hardware and tools, keeping contents clean and protected from moisture.
  • Zipper closure ensures secure sealing and reusability
  • Manufactured from high clarity polyethylene film
  • Meets FDA and USDA specifications for safe contact
  • Thickness of 4 mil provides durability for demanding applications
  • Inside usable dimensions are 12 in W x 15 in H