Learn More
MSC Value Collection Polybags, 3 x 4 in., 2 mil, 1000/Pack
Catalog No. 5000793812
Encompass
These clear polybags are 3 in. in width and 4 in. in height with a 2 mil thickness. Manufactured from 100% virgin, high clarity polyethylene film, they meet FDA and USDA specifications. These bags provide a versatile and flexible storage solution, effectively protecting contents from dirt, dust, and moisture for various applications.
  • Manufactured from 100% virgin, high clarity polyethylene film
  • Meet FDA and USDA specifications for safety
  • Protect contents from dirt, dust, and moisture
  • Features an open top, sealable with twist ties, tape, or impulse sealer
  • Recyclable for environmental consideration
  • Two mil thickness suitable for light to medium weight products
  • Dimensions measured as width x length for inside capacity
Catalog No. 50-007-93812 Supplier MSC Supplier No. 94234895
These clear polybags are 3 in. in width and 4 in. in height with a 2 mil thickness. Manufactured from 100% virgin, high clarity polyethylene film, they meet FDA and USDA specifications. These bags provide a versatile and flexible storage solution, effectively protecting contents from dirt, dust, and moisture for various applications.
  • Manufactured from 100% virgin, high clarity polyethylene film
  • Meet FDA and USDA specifications for safety
  • Protect contents from dirt, dust, and moisture
  • Features an open top, sealable with twist ties, tape, or impulse sealer
  • Recyclable for environmental consideration
  • Two mil thickness suitable for light to medium weight products
  • Dimensions measured as width x length for inside capacity