Learn More
MSC Value Collection Polybags, 4 mil, 4x6 in, Clear, 1000/Pk.
Catalog No. 5000781946
Encompass
Poly bags offer a versatile and flexible storage solution for various items, including food, industrial tools, and electronic equipment. These bags protect contents from dirt, dust, and moisture, and are manufactured from 100% virgin, high-clarity polyethylene film, meeting FDA and USDA specifications.
  • Manufactured from 100% virgin, high clarity polyethylene film
  • Meets FDA and USDA specifications
  • Protects contents from dirt, dust, and moisture
  • Open top can be closed with twist ties, bag tape, or an impulse sealer
  • Recyclable
  • Heavy-duty 4 mil thickness
  • Dimensions represent inside measurements
Catalog No. 50-007-81946 Supplier MSC Supplier No. 12663852
Poly bags offer a versatile and flexible storage solution for various items, including food, industrial tools, and electronic equipment. These bags protect contents from dirt, dust, and moisture, and are manufactured from 100% virgin, high-clarity polyethylene film, meeting FDA and USDA specifications.
  • Manufactured from 100% virgin, high clarity polyethylene film
  • Meets FDA and USDA specifications
  • Protects contents from dirt, dust, and moisture
  • Open top can be closed with twist ties, bag tape, or an impulse sealer
  • Recyclable
  • Heavy-duty 4 mil thickness
  • Dimensions represent inside measurements