Learn More
MSC Value Collection Polyester Twisted Rope, 1/2 in Diameter, 600 ft
Catalog No. 5000532421
Encompass
This polyester twisted rope is provided in a 600 ft total coil, featuring a 1/2 inch diameter and a capacity of 636 pounds. It is suitable for various applications requiring a strong and durable rope.
  • Constructed from durable polyester material.
  • Features a twisted rope design for enhanced strength.
  • Measures 1/2 inch in diameter.
  • Supplied in a 600 foot coil.
  • Provides a capacity of 636 pounds.
  • Available as cut-to-length items in 50 foot multiples.
Catalog No. 50-005-32421 Supplier MSC Supplier No. 45901329
This polyester twisted rope is provided in a 600 ft total coil, featuring a 1/2 inch diameter and a capacity of 636 pounds. It is suitable for various applications requiring a strong and durable rope.
  • Constructed from durable polyester material.
  • Features a twisted rope design for enhanced strength.
  • Measures 1/2 inch in diameter.
  • Supplied in a 600 foot coil.
  • Provides a capacity of 636 pounds.
  • Available as cut-to-length items in 50 foot multiples.