Learn More
MSC Value Collection Polyester Twisted Rope, 1/4 in, 165 lb, 200 ft
Catalog No. 5000514924
Encompass
This polyester twisted rope features a 1/4 inch diameter and a robust 165 lb capacity. It is supplied in convenient 200 ft total coils, making it suitable for a wide range of general-purpose and light-duty applications requiring strength and durability.
  • Made from polyester material
  • Twisted rope construction
  • 1/4 in diameter
  • 165 lb capacity
  • 200 ft total coil length
Catalog No. 50-005-14924 Supplier MSC Supplier No. 45901717
This polyester twisted rope features a 1/4 inch diameter and a robust 165 lb capacity. It is supplied in convenient 200 ft total coils, making it suitable for a wide range of general-purpose and light-duty applications requiring strength and durability.
  • Made from polyester material
  • Twisted rope construction
  • 1/4 in diameter
  • 165 lb capacity
  • 200 ft total coil length