Learn More
MSC Value Collection Polyimide Film Tape, 3/8 in Wide, 1 ea
Catalog No. 5000796133
Encompass
Polyimide film tape from Value Collection is designed for high-temperature applications. It features a silicone adhesive and measures 3/8 in wide by 108 ft long, with a 2.5 mil thickness, providing robust performance across a broad temperature range.
  • Thickness of 2.5 mil
  • Amber color
  • Width of 3/8 in
  • Length of 108 ft
  • Silicone adhesive
  • Polyimide material
  • Tensile strength of 30 lb/in
  • Operating temperature range: -40°F to 500°F
Catalog No. 50-007-96133 Supplier MSC Supplier No. 20717336
Polyimide film tape from Value Collection is designed for high-temperature applications. It features a silicone adhesive and measures 3/8 in wide by 108 ft long, with a 2.5 mil thickness, providing robust performance across a broad temperature range.
  • Thickness of 2.5 mil
  • Amber color
  • Width of 3/8 in
  • Length of 108 ft
  • Silicone adhesive
  • Polyimide material
  • Tensile strength of 30 lb/in
  • Operating temperature range: -40°F to 500°F