Learn More
MSC Value Collection Precision Level, 8 in Length, 0.0005 in Sensitivity Per 10 Inches, Cast Iron, 2 Vials
Catalog No. 5000529316
Encompass
This inspection block precision level is designed for accurate leveling tasks. It features a cast iron construction with two vials, offering high sensitivity for precise measurements over an 8-inch length.
  • Supports business needs with quality products at great value.
  • Constructed from cast iron for durability.
  • Equipped with two vials for accurate readings.
  • Provides a graduation sensitivity of 0.0005 in per 10 inches.
  • Features a base flatness of 0.000400 "
  • Manufactured in Japan.
Catalog No. 50-005-29316 Supplier MSC Supplier No. 06530273
This inspection block precision level is designed for accurate leveling tasks. It features a cast iron construction with two vials, offering high sensitivity for precise measurements over an 8-inch length.
  • Supports business needs with quality products at great value.
  • Constructed from cast iron for durability.
  • Equipped with two vials for accurate readings.
  • Provides a graduation sensitivity of 0.0005 in per 10 inches.
  • Features a base flatness of 0.000400 "
  • Manufactured in Japan.