Learn More
MSC Value Collection Protractor, 6 in Blade, 180° Max Measurement, 1° Dial Graduation
Catalog No. 5000513307
Encompass
This protractor is designed for precise angle measurements in various applications. It features a 6-inch long blade and a rectangular head, allowing for accurate readings up to 180 degrees with a dial graduation of 1 degree.
  • Rectangular head for stable positioning.
  • 6 in blade for measuring various lengths.
  • 180 degree maximum measurement.
  • 1 degree dial graduation for precise readings.
  • Dial range of 0-180 degrees.
  • Manufactured in India.
Catalog No. 50-005-13307 Supplier MSC Supplier No. 06475172
This protractor is designed for precise angle measurements in various applications. It features a 6-inch long blade and a rectangular head, allowing for accurate readings up to 180 degrees with a dial graduation of 1 degree.
  • Rectangular head for stable positioning.
  • 6 in blade for measuring various lengths.
  • 180 degree maximum measurement.
  • 1 degree dial graduation for precise readings.
  • Dial range of 0-180 degrees.
  • Manufactured in India.