The Value Collection PVC Tube is a flexible, clear tubing designed for various applications. It features an inside diameter of 3/8 inch and an outside diameter of 5/8 inch, with a 1/8 inch wall thickness. This tubing is rated for a maximum working pressure of 65 psi and can withstand temperatures ranging from 25 to 150°F. It is FDA compliant and meets REACH, Prop 65, and RoHS standards.
- Flexible design for easy handling and installation.
- Clear tubing allows for visual flow monitoring.
- Withstands a maximum pressure of 65 psi.
- Operates effectively in temperatures from 25 to 150°F.
- FDA compliant for safe use in various applications.
- Meets REACH, Prop 65, and RoHS standards for assured quality.
- Available in a minimum cut length of 25 feet with a standard coil length of 100 feet.