Learn More
MSC Value Collection Radius Gage, 11/64 Inch Radius, Chrome Coated, Steel
Catalog No. 5000529508
Encompass
The Value Collection radius gage is designed for precise measurement of radii. It features an 11/64 inch radius, is chrome coated for durability, and constructed from steel, offering reliable performance for various applications.
  • 11/64 inch radius for accurate measurement.
  • Chrome coated for enhanced durability and corrosion resistance.
  • Steel construction ensures strength and longevity.
  • Suitable for various industrial and workshop applications.
Catalog No. 50-005-29508 Supplier MSC Supplier No. 06474472
The Value Collection radius gage is designed for precise measurement of radii. It features an 11/64 inch radius, is chrome coated for durability, and constructed from steel, offering reliable performance for various applications.
  • 11/64 inch radius for accurate measurement.
  • Chrome coated for enhanced durability and corrosion resistance.
  • Steel construction ensures strength and longevity.
  • Suitable for various industrial and workshop applications.