Learn More
MSC Value Collection Radius Gage Set, Steel, 0.01-0.5 in, 26 pc
Catalog No. 5000530779
Encompass
This radius gage set is designed for precise measurement of radii, featuring a durable steel construction with a satin chrome finish. It includes 26 individual gages to cover a wide range of measurements, making it suitable for various inspection and quality control applications.
  • Durable steel construction.
  • Satin chrome finish for enhanced durability.
  • Comprehensive set of 26 individual gages.
  • Covers a radius range from 0.01 to 0.5 inches.
  • Includes a holder for convenient use.
  • Holder securely grasps gages for precise presentation to the part.
Catalog No. 50-005-30779 Supplier MSC Supplier No. 76457001
This radius gage set is designed for precise measurement of radii, featuring a durable steel construction with a satin chrome finish. It includes 26 individual gages to cover a wide range of measurements, making it suitable for various inspection and quality control applications.
  • Durable steel construction.
  • Satin chrome finish for enhanced durability.
  • Comprehensive set of 26 individual gages.
  • Covers a radius range from 0.01 to 0.5 inches.
  • Includes a holder for convenient use.
  • Holder securely grasps gages for precise presentation to the part.