Learn More
MSC Value Collection Radius Gage Set, Steel, 0.55-1 in Radius, 0.05 in Graduation, 10 pc
Catalog No. 5000530601
Encompass
This radius gage set is constructed from durable steel and features a satin chrome finish, ensuring longevity and ease of readability. The set includes 10 pieces, offering a range of radii from 0.55 to 1 inch with a precise graduation of 0.05 inch, suitable for a variety of measurement applications.
  • Satin chrome finish
  • Constructed from steel
  • Set includes 10 pieces
  • Offers a radius range of 0.55 to 1 inch
  • Features 0.05 inch graduation for precise measurements
Catalog No. 50-005-30601 Supplier MSC Supplier No. 63326938
This radius gage set is constructed from durable steel and features a satin chrome finish, ensuring longevity and ease of readability. The set includes 10 pieces, offering a range of radii from 0.55 to 1 inch with a precise graduation of 0.05 inch, suitable for a variety of measurement applications.
  • Satin chrome finish
  • Constructed from steel
  • Set includes 10 pieces
  • Offers a radius range of 0.55 to 1 inch
  • Features 0.05 inch graduation for precise measurements