Learn More
MSC Value Collection Radius Gage Set, Steel, 1-9/16 to 2 in Radius, 1/16 in Graduation
Catalog No. 5000529182
Encompass
This radius gage set is designed for precise measurement of radii. Made from steel and featuring a satin chrome finish, it includes 8 pieces covering a radius range from 1-9/16 inches to 2 inches with 1/16 inch graduations, offering quality and value for various business needs.
  • Constructed from durable steel for longevity.
  • Features a satin chrome finish for enhanced durability and readability.
  • Measures radii from 1-9/16 inches to 2 inches.
  • Includes 8 individual pieces for comprehensive radius measurement.
  • Provides precise measurements with 1/16 inch graduation.
  • Offers a great value solution for various business measurement requirements.
Catalog No. 50-005-29182 Supplier MSC Supplier No. 06457097
This radius gage set is designed for precise measurement of radii. Made from steel and featuring a satin chrome finish, it includes 8 pieces covering a radius range from 1-9/16 inches to 2 inches with 1/16 inch graduations, offering quality and value for various business needs.
  • Constructed from durable steel for longevity.
  • Features a satin chrome finish for enhanced durability and readability.
  • Measures radii from 1-9/16 inches to 2 inches.
  • Includes 8 individual pieces for comprehensive radius measurement.
  • Provides precise measurements with 1/16 inch graduation.
  • Offers a great value solution for various business measurement requirements.