Learn More
MSC Value Collection Ratcheting Jack Stand, 3 Ton Load Capacity, 11-1/2-17 in Height
Catalog No. 5000511230
Encompass
This ratcheting jack stand offers a 3 ton load capacity, suitable for safely supporting vehicles or heavy objects. It features a height range of 11-1/2 to 17 inches and has a base with dimensions of 6-3/4 inches by 7-3/4 inches, providing stable support. The design includes a ratcheting mechanism for easy height adjustments.
  • Ratcheting jack stand type
  • 3 ton (6000 lb.) load capacity
  • 11-1/2 in minimum height
  • 17 in maximum height
  • 6-3/4 in base width
  • 7-3/4 in base length
  • 7-3/4 in saddle width
  • 6-3/4 in saddle length
  • Country of origin: China
Catalog No. 50-005-11230 Supplier MSC Supplier No. 51374163
This ratcheting jack stand offers a 3 ton load capacity, suitable for safely supporting vehicles or heavy objects. It features a height range of 11-1/2 to 17 inches and has a base with dimensions of 6-3/4 inches by 7-3/4 inches, providing stable support. The design includes a ratcheting mechanism for easy height adjustments.
  • Ratcheting jack stand type
  • 3 ton (6000 lb.) load capacity
  • 11-1/2 in minimum height
  • 17 in maximum height
  • 6-3/4 in base width
  • 7-3/4 in base length
  • 7-3/4 in saddle width
  • 6-3/4 in saddle length
  • Country of origin: China