Learn More
MSC Value Collection Reclosable Polybag, Zipper, 18 x 24 in, 100/Pk.
Catalog No. 5000792643
Encompass
Reclosable polybags, manufactured from 100% virgin, high clarity polyethylene film, meet FDA and USDA specifications. These bags are designed for secure storage, with sizes representing inside usable dimensions.
  • Zipper extends bag width, keeping products clean and free from moisture
  • Bags are reusable
  • Made from 100% virgin, high clarity polyethylene film
  • Meets FDA and USDA specifications
  • Sizes represent inside usable dimensions
  • A 6 mil thickness provides extra heavy-duty puncture and tear resistance
Catalog No. 50-007-92643 Supplier MSC Supplier No. 94235058
Reclosable polybags, manufactured from 100% virgin, high clarity polyethylene film, meet FDA and USDA specifications. These bags are designed for secure storage, with sizes representing inside usable dimensions.
  • Zipper extends bag width, keeping products clean and free from moisture
  • Bags are reusable
  • Made from 100% virgin, high clarity polyethylene film
  • Meets FDA and USDA specifications
  • Sizes represent inside usable dimensions
  • A 6 mil thickness provides extra heavy-duty puncture and tear resistance