Learn More
MSC Value Collection Rectangular Gage Block, 0.21 in, Steel, AS-1 Grade
Catalog No. 5000515084
Encompass
This rectangular gage block from Value Collection is designed for precision measurement and calibration tasks. It offers reliable accuracy for various industrial and laboratory applications.
  • Made from durable steel material
  • Features a 0.21 in size
  • Conforms to AS-1 accuracy grade standards
  • Weighs approximately 0.20 lb for easy handling
Catalog No. 50-005-15084 Supplier MSC Supplier No. 86294105
This rectangular gage block from Value Collection is designed for precision measurement and calibration tasks. It offers reliable accuracy for various industrial and laboratory applications.
  • Made from durable steel material
  • Features a 0.21 in size
  • Conforms to AS-1 accuracy grade standards
  • Weighs approximately 0.20 lb for easy handling