Learn More
MSC Value Collection Rectangular Steel Gage Block, 0.145 in, Grade 0, NIST Traceability Certified
Catalog No. 5000528960
Encompass
This rectangular steel gage block features a precise 0.145 inch size and an accuracy grade of 0. It includes NIST traceability certification, making it suitable for high-precision measurement applications requiring verifiable accuracy.
  • 0.145 inch size.
  • Rectangular steel construction.
  • Accuracy grade 0.
  • Includes NIST traceability certification.
Catalog No. 50-005-28960 Supplier MSC Supplier No. 86283256
This rectangular steel gage block features a precise 0.145 inch size and an accuracy grade of 0. It includes NIST traceability certification, making it suitable for high-precision measurement applications requiring verifiable accuracy.
  • 0.145 inch size.
  • Rectangular steel construction.
  • Accuracy grade 0.
  • Includes NIST traceability certification.