Learn More
MSC Value Collection Rigid Top Plate Caster, Phenolic, 4 in Wheel Dia, 800 lb
Catalog No. 5000529788
Encompass
This rigid top plate caster is designed for general purpose applications, offering robust performance with a phenolic wheel and a durable zinc finish. It features a roller bearing for smooth and reliable movement.
  • Phenolic wheel material
  • 4 inch wheel diameter
  • 2 inch wheel width
  • 800 lb load capacity
  • Roller bearing type
  • Stamp formed construction
  • 4 x 4-1/2 inch plate size
  • 5-5/8 inch overall height
  • Zinc finish
  • General purpose application
  • Top plate mount
  • Rigid style
Catalog No. 50-005-29788 Supplier MSC Supplier No. 02277218
This rigid top plate caster is designed for general purpose applications, offering robust performance with a phenolic wheel and a durable zinc finish. It features a roller bearing for smooth and reliable movement.
  • Phenolic wheel material
  • 4 inch wheel diameter
  • 2 inch wheel width
  • 800 lb load capacity
  • Roller bearing type
  • Stamp formed construction
  • 4 x 4-1/2 inch plate size
  • 5-5/8 inch overall height
  • Zinc finish
  • General purpose application
  • Top plate mount
  • Rigid style