Learn More
MSC Value Collection Rigid Top Plate Caster, Polyurethane, 4 in Dia, 700 lb
Catalog No. 5000531561
Encompass
This rigid top plate caster features a polyurethane wheel with a 4 inch diameter and 2 inch width, providing a load capacity of 700 lb. It has an overall height of 5-5/8 inches and is designed for material handling applications. The caster includes a roller bearing, a 4 x 4-1/2 inch plate, and a zinc finish with stamp formed construction.
  • Polyurethane wheel material (Polyurethane Mold on Steel)
  • 4 inch wheel diameter
  • 2 inch wheel width
  • 700 lb load capacity
  • 5-5/8 inch overall height
  • Roller bearing type
  • 4 x 4-1/2 inch plate size
  • Zinc finish and coating
  • Stamp formed construction
  • Top plate mount
  • Rigid style
Catalog No. 50-005-31561 Supplier MSC Supplier No. 66028150
This rigid top plate caster features a polyurethane wheel with a 4 inch diameter and 2 inch width, providing a load capacity of 700 lb. It has an overall height of 5-5/8 inches and is designed for material handling applications. The caster includes a roller bearing, a 4 x 4-1/2 inch plate, and a zinc finish with stamp formed construction.
  • Polyurethane wheel material (Polyurethane Mold on Steel)
  • 4 inch wheel diameter
  • 2 inch wheel width
  • 700 lb load capacity
  • 5-5/8 inch overall height
  • Roller bearing type
  • 4 x 4-1/2 inch plate size
  • Zinc finish and coating
  • Stamp formed construction
  • Top plate mount
  • Rigid style