Learn More
MSC Value Collection Rigid Top Plate Caster, Rubber, 8 in Wheel Dia, 2 in Wheel Width, 600 lb Capacity
Catalog No. 5000537409
Encompass
This rigid top plate caster features an 8 inch rubber wheel with a 2 inch width, designed for material handling applications. It has a load capacity of 600 lb and an overall height of 9-1/2 inches, making it suitable for various industrial uses.
  • Rigid top plate caster design for stable and secure mounting.
  • Rubber wheel material provides good traction and shock absorption.
  • 8 inch wheel diameter and 2 inch wheel width for smooth movement.
  • Roller bearing type ensures efficient operation.
  • 600 lb load capacity for demanding material handling tasks.
  • Zinc finish/coating for enhanced durability.
  • Stamp formed construction for robust performance.
Catalog No. 50-005-37409 Supplier MSC Supplier No. 66028366
This rigid top plate caster features an 8 inch rubber wheel with a 2 inch width, designed for material handling applications. It has a load capacity of 600 lb and an overall height of 9-1/2 inches, making it suitable for various industrial uses.
  • Rigid top plate caster design for stable and secure mounting.
  • Rubber wheel material provides good traction and shock absorption.
  • 8 inch wheel diameter and 2 inch wheel width for smooth movement.
  • Roller bearing type ensures efficient operation.
  • 600 lb load capacity for demanding material handling tasks.
  • Zinc finish/coating for enhanced durability.
  • Stamp formed construction for robust performance.