Learn More
MSC Value Collection Rockwell A, B, C Bench Top Hardness Tester, Dial Display
Catalog No. 5000527477
Encompass
The Rockwell A, B, C Bench Top Hardness Tester is designed for accurate hardness measurements across multiple Rockwell scales. Featuring a dial display, this tester provides reliable results for various materials.
  • Measures hardness on Rockwell A, B, and C scales.
  • Features a dial display for clear readings.
  • Minimum hardness: 20 RA, 20 RB, 20 RC.
  • Maximum hardness: 100 RB, 70 RC, 88 RA.
  • Manufactured in China.
Catalog No. 50-005-27477 Supplier MSC Supplier No. 06534051
The Rockwell A, B, C Bench Top Hardness Tester is designed for accurate hardness measurements across multiple Rockwell scales. Featuring a dial display, this tester provides reliable results for various materials.
  • Measures hardness on Rockwell A, B, and C scales.
  • Features a dial display for clear readings.
  • Minimum hardness: 20 RA, 20 RB, 20 RC.
  • Maximum hardness: 100 RB, 70 RC, 88 RA.
  • Manufactured in China.