Learn More
MSC Value Collection Roller Chain, 1-1/4 in Pitch, 100R Trade, 10 ft Long
Catalog No. 5000536840
Encompass
This roller chain is designed for various power transmission applications, offering reliable performance. It features a 1-1/4 inch pitch and 100R trade size, provided in a 10-foot length. This single strand chain is built to provide quality and value for industrial needs.
  • Designed for power transmission applications.
  • Provides reliable performance.
  • Features a 1-1/4 inch pitch.
  • Has a 100R trade size.
  • Provided in 10-foot lengths.
  • Single strand configuration.
  • Offers quality and value.
Catalog No. 50-005-36840 Supplier MSC Supplier No. 53590360
This roller chain is designed for various power transmission applications, offering reliable performance. It features a 1-1/4 inch pitch and 100R trade size, provided in a 10-foot length. This single strand chain is built to provide quality and value for industrial needs.
  • Designed for power transmission applications.
  • Provides reliable performance.
  • Features a 1-1/4 inch pitch.
  • Has a 100R trade size.
  • Provided in 10-foot lengths.
  • Single strand configuration.
  • Offers quality and value.