Learn More
MSC Value Collection Roller Chain, 1 in Pitch, 80R Trade, 10 ft
Catalog No. 5000527085
Encompass
This roller chain is designed for various business needs, offering a 1 inch pitch, 80R trade size, and a length of 10 feet. It also features a 0.625 inch roller diameter.
  • Single strand chain style.
  • 1 in chain pitch.
  • ANSI number 80.
  • 80R chain trade size.
  • 0.625 in roller diameter.
  • 0.6250 in roller width.
  • 10 ft length.
  • 0.3120 in pin width.
Catalog No. 50-005-27085 Supplier MSC Supplier No. 53590402
This roller chain is designed for various business needs, offering a 1 inch pitch, 80R trade size, and a length of 10 feet. It also features a 0.625 inch roller diameter.
  • Single strand chain style.
  • 1 in chain pitch.
  • ANSI number 80.
  • 80R chain trade size.
  • 0.625 in roller diameter.
  • 0.6250 in roller width.
  • 10 ft length.
  • 0.3120 in pin width.