Learn More
MSC Value Collection Roller Chain, 5/8 in Pitch, ANSI 50, Single Strand
Catalog No. 5000512909
Encompass
This roller chain offers a reliable and cost-effective solution for various industrial applications. Designed for durability and performance, it features a single strand configuration and adheres to industry-standard specifications for consistent operation.
  • Single strand style
  • 5/8 inch pitch
  • ANSI 50 compliant
  • ISO 10A-1 compliant
  • Roller diameter of 0.400 inch
  • Roller width of 0.3750 inch
  • Pin width of 0.2000 inch
  • Provides great value to support business needs
Catalog No. 50-005-12909 Supplier MSC Supplier No. 53614087
This roller chain offers a reliable and cost-effective solution for various industrial applications. Designed for durability and performance, it features a single strand configuration and adheres to industry-standard specifications for consistent operation.
  • Single strand style
  • 5/8 inch pitch
  • ANSI 50 compliant
  • ISO 10A-1 compliant
  • Roller diameter of 0.400 inch
  • Roller width of 0.3750 inch
  • Pin width of 0.2000 inch
  • Provides great value to support business needs