Learn More
MSC Value Collection Rope, Polypropylene, High Strength, 1/2 in, 762 Lb
Catalog No. 5000535184
Encompass
This polypropylene high strength rope is sold in 1 ft. increments and is available as a 200 ft. total coil. It offers a robust capacity of 762 Lb and has a diameter of 1/2 in, making it suitable for demanding applications requiring strong and durable cordage.
  • Capacity of 762 Lb.
  • Diameter of 1/2 in.
  • Made from polypropylene material.
  • High strength rope type.
  • Sold in 1 ft. increments.
  • Available as a 200 ft. total coil.
  • Manufactured in the United States.
Catalog No. 50-005-35184 Supplier MSC Supplier No. 45900362
This polypropylene high strength rope is sold in 1 ft. increments and is available as a 200 ft. total coil. It offers a robust capacity of 762 Lb and has a diameter of 1/2 in, making it suitable for demanding applications requiring strong and durable cordage.
  • Capacity of 762 Lb.
  • Diameter of 1/2 in.
  • Made from polypropylene material.
  • High strength rope type.
  • Sold in 1 ft. increments.
  • Available as a 200 ft. total coil.
  • Manufactured in the United States.