Learn More
MSC Value Collection Safety Glasses, Anti-Fog, Polycarbonate, Full-Framed, 1 ea
Catalog No. 5000793706
Encompass
These safety glasses are designed for enhanced visibility in hot and humid conditions. They feature clear polycarbonate lenses, a full-framed design, and a gray frame with yellow temples.
  • Anti-fog coating
  • Clear polycarbonate lenses
  • Full-framed design
  • Gray frame, yellow temples
  • Integrated nose pad
  • UV protection
  • Compliant with ANSI Z87.1-2010
  • Non-adjustable temples
Catalog No. 50-007-93706 Supplier MSC Supplier No. 92590520
These safety glasses are designed for enhanced visibility in hot and humid conditions. They feature clear polycarbonate lenses, a full-framed design, and a gray frame with yellow temples.
  • Anti-fog coating
  • Clear polycarbonate lenses
  • Full-framed design
  • Gray frame, yellow temples
  • Integrated nose pad
  • UV protection
  • Compliant with ANSI Z87.1-2010
  • Non-adjustable temples