Learn More
MSC Value Collection Shortway Reducer, 8/7 in ID, Galvanized, 1 ea
Catalog No. 5000504381
Encompass
This galvanized duct shortway reducer is designed without a crimp, providing a transition between ductwork sections. It features an 8-inch inside diameter reducing to a 7-inch inside diameter, suitable for various ventilation and exhaust applications.
  • Duct fitting type: Shortway reducer wo/crimp
  • Inside diameter: 8 in
  • Material: Galvanized
  • Reduced inside diameter: 7 in
  • Standard gage
Catalog No. 50-005-04381 Supplier MSC Supplier No. 03379468
This galvanized duct shortway reducer is designed without a crimp, providing a transition between ductwork sections. It features an 8-inch inside diameter reducing to a 7-inch inside diameter, suitable for various ventilation and exhaust applications.
  • Duct fitting type: Shortway reducer wo/crimp
  • Inside diameter: 8 in
  • Material: Galvanized
  • Reduced inside diameter: 7 in
  • Standard gage