Learn More
MSC Value Collection Side Mount Pulley, 1-3/8 in OD, Malleable Iron, Galvanized
Catalog No. 5000530634
Encompass
This single side side mount guidance pulley is a sheave designed for fibrous ropes. It features a 1-3/8 in outside diameter and is constructed from malleable iron with a galvanized finish, making it suitable for various applications.
  • Designed for fibrous rope
  • 1-3/8 in outside diameter
  • 5/16 in rope diameter
  • Mounting hole diameter ranges from 0.183 in to 0.194 in
  • Constructed from malleable iron
  • Galvanized finish
Catalog No. 50-005-30634 Supplier MSC Supplier No. 67328609
This single side side mount guidance pulley is a sheave designed for fibrous ropes. It features a 1-3/8 in outside diameter and is constructed from malleable iron with a galvanized finish, making it suitable for various applications.
  • Designed for fibrous rope
  • 1-3/8 in outside diameter
  • 5/16 in rope diameter
  • Mounting hole diameter ranges from 0.183 in to 0.194 in
  • Constructed from malleable iron
  • Galvanized finish