Learn More
MSC Value Collection Side Release Snatch Block, 17,600 lb, Steel, 6 in.
Catalog No. 5000535465
Encompass
This side release snatch block features a single sheave with a 6-inch outside diameter. Constructed from steel with a painted finish, it is designed for heavy-duty applications and accommodates wire ropes of 5/8 inch and 3/4 inch diameters.
  • Load limit of 17,600 lb.
  • Single sheave.
  • 6 in. outside diameter.
  • Accommodates 5/8 in. and 3/4 in. wire rope.
  • Steel construction.
  • Painted finish.
Catalog No. 50-005-35465 Supplier MSC Supplier No. 67327007
This side release snatch block features a single sheave with a 6-inch outside diameter. Constructed from steel with a painted finish, it is designed for heavy-duty applications and accommodates wire ropes of 5/8 inch and 3/4 inch diameters.
  • Load limit of 17,600 lb.
  • Single sheave.
  • 6 in. outside diameter.
  • Accommodates 5/8 in. and 3/4 in. wire rope.
  • Steel construction.
  • Painted finish.