Learn More
MSC Value Collection Single Closed Upright Mount Guidance Pulley, Iron, Galvanized, 3 in OD
Catalog No. 5000536085
Encompass
This guidance pulley is part of the Value Collection, offering quality products for your business needs. It features a single closed upright mount, constructed from galvanized iron, and has a 3 in outside diameter.
  • Sheave, 3 in outside diameter
  • Designed for 3/8 in diameter wire rope
  • Mounting hole diameter 0.235 in to 0.246 in
  • Iron construction with galvanized finish
  • Quality product at a great value
Catalog No. 50-005-36085 Supplier MSC Supplier No. 67328021
This guidance pulley is part of the Value Collection, offering quality products for your business needs. It features a single closed upright mount, constructed from galvanized iron, and has a 3 in outside diameter.
  • Sheave, 3 in outside diameter
  • Designed for 3/8 in diameter wire rope
  • Mounting hole diameter 0.235 in to 0.246 in
  • Iron construction with galvanized finish
  • Quality product at a great value