Learn More
MSC Value Collection Single Jack Chain, 0.134 in, 43 lb, Zinc Plated
Catalog No. 5000512126
Encompass
This single jack chain is designed for various applications, featuring a 0.134 in diameter and a work load limit of 43 lb. It is constructed from steel with a zinc plated finish for durability. The chain has an inside length of 1.04 in and is supplied in a 100 ft coil.
  • Constructed from steel
  • Zinc plated finish
  • 0.134 in diameter
  • 43 lb work load limit
  • 1.04 in inside length
  • 100 ft total coil
Catalog No. 50-005-12126 Supplier MSC Supplier No. 66795535
This single jack chain is designed for various applications, featuring a 0.134 in diameter and a work load limit of 43 lb. It is constructed from steel with a zinc plated finish for durability. The chain has an inside length of 1.04 in and is supplied in a 100 ft coil.
  • Constructed from steel
  • Zinc plated finish
  • 0.134 in diameter
  • 43 lb work load limit
  • 1.04 in inside length
  • 100 ft total coil