Learn More
MSC Value Collection Sisal Twine Tube, 2 Ply, 312 lb Breaking Strength
Catalog No. 5000527698
Encompass
This sisal twine is designed for various applications, offering a high breaking strength and a natural color. It comes in a convenient tube format, providing reliability and durability for your needs.
  • Twine made from durable sisal material.
  • Natural color for versatile use.
  • Features a 2 ply construction.
  • Offers a breaking strength of 312 lb.
  • Provides 147 ft per pound.
  • Supplied as a 147 foot roll.
Catalog No. 50-005-27698 Supplier MSC Supplier No. 45890811
This sisal twine is designed for various applications, offering a high breaking strength and a natural color. It comes in a convenient tube format, providing reliability and durability for your needs.
  • Twine made from durable sisal material.
  • Natural color for versatile use.
  • Features a 2 ply construction.
  • Offers a breaking strength of 312 lb.
  • Provides 147 ft per pound.
  • Supplied as a 147 foot roll.