Learn More
MSC Value Collection Slotted Spring Pin, 4 mm, 30 mm, 1070-1080 Steel
Catalog No. 5000509034
Encompass
This slotted spring pin is made from 1070-1080 steel with a plain finish, featuring a 4 mm diameter and 30 mm overall length. It is designed for metric system applications.
  • Slotted pin type
  • 30 mm overall length
  • 4 mm diameter
  • Made from 1070-1080 steel
  • Plain finish
  • Metric system of measurement
Catalog No. 50-005-09034 Supplier MSC Supplier No. 01048891
This slotted spring pin is made from 1070-1080 steel with a plain finish, featuring a 4 mm diameter and 30 mm overall length. It is designed for metric system applications.
  • Slotted pin type
  • 30 mm overall length
  • 4 mm diameter
  • Made from 1070-1080 steel
  • Plain finish
  • Metric system of measurement