Learn More
MSC Value Collection Slotted Spring Pin, 6 mm x 18 mm, 1070-1080 Steel
Catalog No. 5000507995
Encompass
This slotted spring pin is manufactured from 1070-1080 steel with a plain finish, designed for various industrial and mechanical fastening applications.
  • Slotted pin type
  • Constructed from 1070-1080 steel
  • Plain finish
  • Metric system of measurement
  • 6 mm diameter
  • 18 mm overall length
  • Made in the United Kingdom
Catalog No. 50-005-07995 Supplier MSC Supplier No. 01049238
This slotted spring pin is manufactured from 1070-1080 steel with a plain finish, designed for various industrial and mechanical fastening applications.
  • Slotted pin type
  • Constructed from 1070-1080 steel
  • Plain finish
  • Metric system of measurement
  • 6 mm diameter
  • 18 mm overall length
  • Made in the United Kingdom