Learn More
MSC Value Collection Square Steel Gage Block, 0.108 in, Grade 0
Catalog No. 5000535618
Encompass
This square steel gage block is precisely crafted to 0.108 inch and features an accuracy grade of 0. It includes NIST certification, ensuring high accuracy and reliability for various measurement applications in industrial and laboratory settings.
  • Includes NIST certification
  • Constructed from durable steel
  • Features a precise square shape
  • Manufactured to accuracy grade 0
  • Measures 0.108 inch in size
Catalog No. 50-005-35618 Supplier MSC Supplier No. 86271459
This square steel gage block is precisely crafted to 0.108 inch and features an accuracy grade of 0. It includes NIST certification, ensuring high accuracy and reliability for various measurement applications in industrial and laboratory settings.
  • Includes NIST certification
  • Constructed from durable steel
  • Features a precise square shape
  • Manufactured to accuracy grade 0
  • Measures 0.108 inch in size