Learn More
MSC Value Collection Square Steel Gage Block, 0.123 in, Grade 0
Catalog No. 5000510611
Encompass
This gage block is designed for precise measurement applications. Made from steel, it features a square shape and provides a size of 0.123 inch with Grade 0 accuracy.
  • NIST certification included
  • Grade 0 accuracy
  • 0.123 inch size
  • Steel material
Catalog No. 50-005-10611 Supplier MSC Supplier No. 86272150
This gage block is designed for precise measurement applications. Made from steel, it features a square shape and provides a size of 0.123 inch with Grade 0 accuracy.
  • NIST certification included
  • Grade 0 accuracy
  • 0.123 inch size
  • Steel material