Learn More
MSC Value Collection Square Steel Gage Block, 0.128 Inch, Grade 0
Catalog No. 5000510612
Encompass
This square steel gage block from Value Collection is designed for precision measurement, featuring an accuracy grade of 0. Each block measures 0.128 inch and comes with a NIST traceability certification.
  • Constructed from steel for durability
  • Dimensions of 0.128 inch
  • Accuracy grade of 0
  • Supplied with NIST traceability certification
Catalog No. 50-005-10612 Supplier MSC Supplier No. 86272408
This square steel gage block from Value Collection is designed for precision measurement, featuring an accuracy grade of 0. Each block measures 0.128 inch and comes with a NIST traceability certification.
  • Constructed from steel for durability
  • Dimensions of 0.128 inch
  • Accuracy grade of 0
  • Supplied with NIST traceability certification