Learn More
MSC Value Collection Square Steel Gage Block, 2 in, Grade 0
Catalog No. 5000510736
Encompass
This square steel gage block measures 2 inches and features an accuracy grade of 0. Constructed from durable steel, it includes NIST certification, ensuring high precision for various applications.
  • Square shape
  • Accuracy grade 0
  • Material: Steel
  • Size: 2 inches
  • Includes NIST certification
Catalog No. 50-005-10736 Supplier MSC Supplier No. 86275807
This square steel gage block measures 2 inches and features an accuracy grade of 0. Constructed from durable steel, it includes NIST certification, ensuring high precision for various applications.
  • Square shape
  • Accuracy grade 0
  • Material: Steel
  • Size: 2 inches
  • Includes NIST certification