Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Value Collection Stand Housing, Use with Worksmart, Gray, 34-1/2 in Height

Catalog No. 5000534670
Encompass

Value Collection offers quality products at a great value to support your business needs.

  • Accessory type is stand housing.
  • For use with compatible tool storage systems.
  • Color is gray.
  • Height is 34-1/2 in.

Catalog No. 50-005-34670 Supplier MSC Supplier No. 68633494
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Value Collection offers quality products at a great value to support your business needs.

  • Accessory type is stand housing.
  • For use with compatible tool storage systems.
  • Color is gray.
  • Height is 34-1/2 in.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.