Learn More
MSC Value Collection Stationary Industrial Work Bench, Static Dissipative, 72 x 36 in, 3600 lb Capacity
Catalog No. 5000516178
Encompass
This industrial work bench is designed for demanding environments, featuring a static dissipative top and body material suitable for ESD-sensitive applications. It offers a robust construction with a high load capacity, making it ideal for various industrial tasks.
  • Industrial work bench
  • 3600 lb load capacity
  • Static dissipative top
  • 72 in overall width
  • 36 in overall depth
  • 34-3/4 in overall height
  • Adjustable leg style
  • Static dissipative body material
  • 1-1/4 in top thickness
Catalog No. 50-005-16178 Supplier MSC Supplier No. 91683383
This industrial work bench is designed for demanding environments, featuring a static dissipative top and body material suitable for ESD-sensitive applications. It offers a robust construction with a high load capacity, making it ideal for various industrial tasks.
  • Industrial work bench
  • 3600 lb load capacity
  • Static dissipative top
  • 72 in overall width
  • 36 in overall depth
  • 34-3/4 in overall height
  • Adjustable leg style
  • Static dissipative body material
  • 1-1/4 in top thickness