Learn More
MSC Value Collection Steel Locking & Storage Cabinet, Gray, 2 Shelf
Catalog No. 5000532592
Encompass
This steel locking and storage cabinet is designed for secure storage in professional environments. With generous dimensions of 36 inches wide, 18 inches deep, and 42 inches high, it provides ample space for organizing various items.
  • Provides both storage and locking capabilities.
  • Constructed from durable steel.
  • Features dimensions of 36 inches wide, 18 inches deep, and 42 inches high.
  • Includes 2 adjustable shelves for flexible organization.
  • Comes in a gray finish.
  • Made with 24 gauge steel for robust construction.
  • Ships unassembled for convenience.
Catalog No. 50-005-32592 Supplier MSC Supplier No. 29286903
This steel locking and storage cabinet is designed for secure storage in professional environments. With generous dimensions of 36 inches wide, 18 inches deep, and 42 inches high, it provides ample space for organizing various items.
  • Provides both storage and locking capabilities.
  • Constructed from durable steel.
  • Features dimensions of 36 inches wide, 18 inches deep, and 42 inches high.
  • Includes 2 adjustable shelves for flexible organization.
  • Comes in a gray finish.
  • Made with 24 gauge steel for robust construction.
  • Ships unassembled for convenience.