Learn More
MSC Value Collection Surface Gage, 3.25 in Base Length, 2 Spindles
Catalog No. 5000513288
Encompass
Value Collection surface gages are reliable tools for precise layout and inspection in various industrial applications. Designed for accuracy, these gages are built to offer dependable performance and support your business needs with quality at a great value.
  • Base length of 3.25 inches
  • Base width of 2.5 inches
  • Two spindles for enhanced stability
  • Supports accurate measurement and layout tasks
Catalog No. 50-005-13288 Supplier MSC Supplier No. 00227371
Value Collection surface gages are reliable tools for precise layout and inspection in various industrial applications. Designed for accuracy, these gages are built to offer dependable performance and support your business needs with quality at a great value.
  • Base length of 3.25 inches
  • Base width of 2.5 inches
  • Two spindles for enhanced stability
  • Supports accurate measurement and layout tasks