Learn More
MSC Value Collection Swivel Top Plate Caster, Phenolic, 5 in Wheel Dia, 1,000 lb Capacity
Catalog No. 5000514676
Encompass
This swivel top plate caster features a durable phenolic wheel and is designed for general purpose applications. It offers a 1,000 lb load capacity and includes a side brake for secure positioning. The caster is constructed with a stamp formed design and finished with zinc plating for added durability.
  • Phenolic wheel material
  • 5 in wheel diameter
  • 2 in wheel width
  • 1,000 lb load capacity
  • Side brake for secure positioning
  • Zinc-plated finish
  • Roller bearing type
  • Stamp formed construction
  • 4 x 4-1/2 in plate size
  • 6-1/2 in overall height
Catalog No. 50-005-14676 Supplier MSC Supplier No. 02276210
This swivel top plate caster features a durable phenolic wheel and is designed for general purpose applications. It offers a 1,000 lb load capacity and includes a side brake for secure positioning. The caster is constructed with a stamp formed design and finished with zinc plating for added durability.
  • Phenolic wheel material
  • 5 in wheel diameter
  • 2 in wheel width
  • 1,000 lb load capacity
  • Side brake for secure positioning
  • Zinc-plated finish
  • Roller bearing type
  • Stamp formed construction
  • 4 x 4-1/2 in plate size
  • 6-1/2 in overall height