Learn More
MSC Value Collection Swivel Top Plate Caster, Phenolic, 8 in Wheel, 900 lb Capacity
Catalog No. 5000514085
Encompass
This swivel top plate caster features a phenolic wheel and is designed for material handling applications. It offers reliable performance with a roller bearing and stamp-formed construction, making it suitable for various industrial uses.
  • Features a roller bearing.
  • Constructed with stamp-formed components.
  • Equipped with a 4 × 4-1/2 in plate size.
  • Finished with a durable zinc coating.
  • Designed for top plate mounting.
  • Allows for swivel movement.
  • Ideal for material handling applications.
Catalog No. 50-005-14085 Supplier MSC Supplier No. 66028069
This swivel top plate caster features a phenolic wheel and is designed for material handling applications. It offers reliable performance with a roller bearing and stamp-formed construction, making it suitable for various industrial uses.
  • Features a roller bearing.
  • Constructed with stamp-formed components.
  • Equipped with a 4 × 4-1/2 in plate size.
  • Finished with a durable zinc coating.
  • Designed for top plate mounting.
  • Allows for swivel movement.
  • Ideal for material handling applications.