Learn More
MSC Value Collection Swivel Top Plate Caster, Polyolefin, 8 in, 1000 lb Capacity
Catalog No. 5000528126
Encompass
This swivel top plate caster is designed to offer quality and value for various material handling applications. It features a durable polyolefin wheel with an 8 inch diameter and 2 inch width, providing smooth movement. Equipped with a roller bearing and a 4 x 4-1/2 inch plate, this caster has a robust 1000 lb load capacity and an overall height of 9-1/2 inches.
  • Polyolefin wheel material
  • 8 inch wheel diameter
  • 2 inch wheel width
  • 1000 lb load capacity
  • 9-1/2 inch overall height
  • Roller bearing type
  • Stamp formed construction
  • 4 x 4-1/2 inch plate size
  • Top plate mount
  • Swivel style
Catalog No. 50-005-28126 Supplier MSC Supplier No. 66028085
This swivel top plate caster is designed to offer quality and value for various material handling applications. It features a durable polyolefin wheel with an 8 inch diameter and 2 inch width, providing smooth movement. Equipped with a roller bearing and a 4 x 4-1/2 inch plate, this caster has a robust 1000 lb load capacity and an overall height of 9-1/2 inches.
  • Polyolefin wheel material
  • 8 inch wheel diameter
  • 2 inch wheel width
  • 1000 lb load capacity
  • 9-1/2 inch overall height
  • Roller bearing type
  • Stamp formed construction
  • 4 x 4-1/2 inch plate size
  • Top plate mount
  • Swivel style