Learn More
MSC Value Collection Swivel Top Plate Caster, Polyurethane, 4 in, 600 lb
Catalog No. 5000512370
Encompass
This swivel top plate caster is designed for general purpose applications, offering reliable mobility. It features a durable polyurethane wheel, a roller bearing, and a robust 600 lb load capacity. Constructed with a stamp formed design and a powder coated finish, it ensures long-lasting performance.
  • Polyurethane wheel material for durability and smooth operation
  • 600 lb load capacity for heavy-duty applications
  • Roller bearing for reduced friction and easy movement
  • Plate size of 4 x 4-1/2 in for secure mounting
  • Wheel diameter of 4 in
  • Wheel width of 2 in
  • Overall height of 5-5/8 in
  • Stamp formed construction for strength
  • Powder coated finish for corrosion resistance
  • Designed for general purpose applications
  • Top plate mount for easy installation
  • Swivel style for versatile maneuverability
Catalog No. 50-005-12370 Supplier MSC Supplier No. 02276962
This swivel top plate caster is designed for general purpose applications, offering reliable mobility. It features a durable polyurethane wheel, a roller bearing, and a robust 600 lb load capacity. Constructed with a stamp formed design and a powder coated finish, it ensures long-lasting performance.
  • Polyurethane wheel material for durability and smooth operation
  • 600 lb load capacity for heavy-duty applications
  • Roller bearing for reduced friction and easy movement
  • Plate size of 4 x 4-1/2 in for secure mounting
  • Wheel diameter of 4 in
  • Wheel width of 2 in
  • Overall height of 5-5/8 in
  • Stamp formed construction for strength
  • Powder coated finish for corrosion resistance
  • Designed for general purpose applications
  • Top plate mount for easy installation
  • Swivel style for versatile maneuverability