Learn More
MSC VALUE COLLECTION Swivel Top Plate Caster, Soft Rubber, 3 inch Wheel Diameter, 200 lb Capacity
Catalog No. 5000514675
Encompass
This swivel top plate caster features a soft rubber wheel, designed for general purpose applications. It offers reliable mobility with a 200 lb load capacity, making it suitable for various needs.
  • Soft rubber wheel material
  • 3 inch wheel diameter
  • 1-1/4 inch wheel width
  • 200 lb load capacity
  • 4-3/8 inch overall height
  • Ball bearing type
  • Top plate mount
  • Stamp formed construction
  • Zinc finish
Catalog No. 50-005-14675 Supplier MSC Supplier No. 02275659
This swivel top plate caster features a soft rubber wheel, designed for general purpose applications. It offers reliable mobility with a 200 lb load capacity, making it suitable for various needs.
  • Soft rubber wheel material
  • 3 inch wheel diameter
  • 1-1/4 inch wheel width
  • 200 lb load capacity
  • 4-3/8 inch overall height
  • Ball bearing type
  • Top plate mount
  • Stamp formed construction
  • Zinc finish